missing translation for 'onlineSavingsMsg'
Learn More

Tripropylene glycol, 99%, pure, mixture of isomers, Thermo Scientific Chemicals

Catalog Number p-7077733
Click to view available options
Quantity:
1 L
5 L
25 L
This item is not returnable. View return policy
24800-44-0
C9H20O4
192.25
This item is not returnable. View return policy
24800-44-0
C9H20O4
192.25

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.0200g/mL
Boiling Point 270.0°C
Flash Point 145°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (sum of isomers) (GC)
Quantity 1 L
Specific Gravity 1.02
Solubility Information Solubility in water: soluble.
Viscosity 77.3 mm²/s (20°C)
Formula Weight 192.25
Percent Purity 99%
Grade Pure
Chemical Name or Material Tripropylene glycol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'