missing translation for 'onlineSavingsMsg'
Learn More

Triisopropanolamine, 95%, Thermo Scientific Chemicals

Catalog Number p-7024765
Click to view available options
Quantity:
10,000 g
2500 g
500 g
This item is not returnable. View return policy
122-20-3
C9H21NO3
191.271
MFCD00004533
SLINHMUFWFWBMU-UHFFFAOYSA-N
triisopropanolamine, tipa, tri-2-propanolamine, tri-iso-propanolamine, tris 2-propanol amine, 2-propanol, 1,1',1-nitrilotris, tris 2-hydroxypropyl amine, 1,1',1-nitrilotripropan-2-ol, tris 2-hydroxy-1-propyl amine, caswell no. 891
24730
1-[bis(2-hydroxypropyl)amino]propan-2-ol
CC(CN(CC(C)O)CC(C)O)O
This item is not returnable. View return policy
122-20-3
C9H21NO3
191.271
MFCD00004533
SLINHMUFWFWBMU-UHFFFAOYSA-N
triisopropanolamine, tipa, tri-2-propanolamine, tri-iso-propanolamine, tris 2-propanol amine, 2-propanol, 1,1',1-nitrilotris, tris 2-hydroxypropyl amine, 1,1',1-nitrilotripropan-2-ol, tris 2-hydroxy-1-propyl amine, caswell no. 891
24730
1-[bis(2-hydroxypropyl)amino]propan-2-ol
CC(CN(CC(C)O)CC(C)O)O

Major applications include water-based coating applications and agricultural products. Additional applications are antistat agents for polymers, corrosion inhibitor, electrodeposition/electrocoating, lubricants, paper, pigment dispersion, plastics, polyurethane additive, reaction intermediates, rubber curing, surfactants, mineral dispersion, and urethanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Major applications include water-based coating applications and agricultural products. Additional applications are antistat agents for polymers, corrosion inhibitor, electrodeposition/electrocoating, lubricants, paper, pigment dispersion, plastics, polyurethane additive, reaction intermediates, rubber curing, surfactants, mineral dispersion, and urethanes.

Solubility
Insoluble in water. Soluble ethanol, diethyl ether, chloroform (slightly ), and methanol (>500 g/100g).

Notes
Hygroscopic. Store away from oxidizing agents, water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY
Melting Point 48°C to 52°C
Boiling Point 189°C to 191°C (23 mmHg)
Flash Point >110°C (230°F)
Odor Amine-like
Linear Formula (CH3CHOHCH2)3N
Quantity 500 g
Beilstein 1071570
Sensitivity Hygroscopic
Solubility Information Insoluble in water. Soluble ethanol,diethyl ether,chloroform (slightly ),and methanol (>500g/100g).
Formula Weight 191.27
Percent Purity 95%
Chemical Name or Material Triisopropanolamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'