Learn More
Uvex Trend Black/Blue PU/PU 69397 Low Shoe - Width 10
Uvex 2 Trend safety footwear for medium-risk applications (S1-S3); heel cap offers increased stability without limiting movement
Brand:  Uvex 6939741
Additional Details : Weight : 1.05300kg
 View more versions of this product
Product Code. 17213941
  • 98.70€ / 1 pair
Estimated Shipment: 24-05-2024
to see stock.
Add to basket
Description
Description
Uvex 2 Trend Shoe 69397 S2 SRC
  • Sporty S2 safety shoe with wide fit
  • Breathable, water repellent microvelour
  • Made from synthetic materials, suitable for people allergic to chrome
  • All sole materials free of silicones, plasticizers and other paint wetting impairment substances
  • Meets ESD specification with volume resistance under 35 megohms
  • Reliable protection for wearers
  • Steel toecap and stabilizing Uvex anti-twist heel cap to prevent ankle twisting
  • Ergonomically designed outsole comprising dual-density polyurethane with excellent slip resistance
  • Reflective details for enhanced visibility
  • Uvex Climazone technology lining, upper and removable comfortable insole
    • Moisture transport system helps maintain foot temperature
  • Soft padding on collar and dust tongue
  • Removable anti-static comfortable insole with a moisture transport system and additional shock absorption around heel and forefoot
  • Article No. 69397
  • Standard EN ISO 20345:2011 S2 SRC
  • Sizes: 38 to 52
Specifications
Specifications
Black, Blue
41
Steel
EN ISO 20345:2011
Narrow width
Yes
S2
Microvelour
Dual-density polyurethane (PU/PU)
No
Unisex
Uvex 2
No
SRC