Learn More
enQuireBio™ TranslationBlocker Mouse TNF alpha siRNA
Blocks transcription and translation of TNF alpha from Mouse.
Brand: enQuireBio™ QX23-10nmol
Additional Details : Weight : 0.01000kg
Specifications
NP_038721 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
21926 | |
10 nmol | |
Mouse |
Murine Peritoneal Macrophage. | |
Tnf | |
siRNA | |
DIF, TNFA, TNFSF2, TNLG1F, TNF-alpha, Tumor necrosis factor alpha, Tumor necrosis factor | |
Lyophilized |