missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human mTOR siRNA
Blocks transcription and translation of mTOR from Human.
250.00€ - 369.00€
Specifications
Accession Number | NP_004949 |
---|---|
Cell Lines Tested | U87MG Human Glioblastoma Cells. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | MTOR |
Gene ID (Entrez) | 2475 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16039251
|
enQuireBio™
QX50-2NMOL |
2 nmol |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16029251
|
enQuireBio™
QX50-10NMOL |
10 nmol |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_004949 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
2475 | |
SKS, FRAP1, RAFT1, FRAP, RAPT1, FRAP2, Serine/threonine-protein kinase mTor, mechanistic target of rapamycin | |
Lyophilized |
U87MG Human Glioblastoma Cells. | |
MTOR | |
siRNA | |
Human |