missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human Caspase-6 siRNA
Blocks transcription and translation of Caspase-6 from Human.
250.00€ - 369.00€
Specifications
Accession Number | NP_001217 |
---|---|
Cell Lines Tested | 293 Human Embryonic Kidney Cells with NF-kappa B and Flag tagged DR4 constructs. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | CASP6 |
Gene ID (Entrez) | 839 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16039231
|
enQuireBio™
QX41-2NMOL |
2 nmol |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16029231
|
enQuireBio™
QX41-10NMOL |
10 nmol |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001217 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
839 | |
MCH2, CASP6 | |
Lyophilized |
293 Human Embryonic Kidney Cells with NF-kappa B and Flag tagged DR4 constructs. | |
CASP6 | |
siRNA | |
Human |