missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human Apoptosis regulator BAX siRNA
Blocks transcription and translation of Apoptosis regulator BAX from Human.
250.00€ - 369.00€
Specifications
Accession Number | NP_004315 |
---|---|
Cell Lines Tested | KAT-4 Human Thyroid Carcinoma Cells and Human T Cells. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | BAX |
Gene ID (Entrez) | 581 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16079251
|
enQuireBio™
QX6-2NMOL |
2 nmol |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16069251
|
enQuireBio™
QX6-10NMOL |
10 nmol |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_004315 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
581 | |
BCL2L4 | |
Lyophilized |
KAT-4 Human Thyroid Carcinoma Cells and Human T Cells. | |
BAX | |
siRNA | |
Human |