missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Ethyl-4'-(4-n-pentylcyclohexyl)biphenyl, 97%, Thermo Scientific™

Catalog Number p-7037076
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
79709-85-6
C25H34
334.55
MFCD01941072
JOLGXBQYTARJLD-UHFFFAOYSA-N
trans-4-ethyl-4'-4-pentylcyclohexyl-1,1'-biphenyl, 1-ethyl-4-4-4-pentylcyclohexyl phenyl benzene, 4-ethyl-4'-trans-4-pentylcyclohexyl-1,1'-biphenyl, 1-4-ethylphenyl-4-4-pentylcyclohexyl benzene, 1,1'-biphenyl, 4-ethyl-4'-4-pentylcyclohexyl, bch-52, bpch-52, 4-ethyl-4'-4-pentylcyclohexyl biphenyl, 4-ethyl-4'-4-pentylcyclohexyl-1,1'-biphenyl, trans-4-ethyl-4'-4-n-pentylcyclohexyl biphenyl
630553
CCCCCC1CCC(CC1)C1=CC=C(C=C1)C1=CC=C(CC)C=C1
This item is not returnable. View return policy
79709-85-6
C25H34
334.55
MFCD01941072
JOLGXBQYTARJLD-UHFFFAOYSA-N
trans-4-ethyl-4'-4-pentylcyclohexyl-1,1'-biphenyl, 1-ethyl-4-4-4-pentylcyclohexyl phenyl benzene, 4-ethyl-4'-trans-4-pentylcyclohexyl-1,1'-biphenyl, 1-4-ethylphenyl-4-4-pentylcyclohexyl benzene, 1,1'-biphenyl, 4-ethyl-4'-4-pentylcyclohexyl, bch-52, bpch-52, 4-ethyl-4'-4-pentylcyclohexyl biphenyl, 4-ethyl-4'-4-pentylcyclohexyl-1,1'-biphenyl, trans-4-ethyl-4'-4-n-pentylcyclohexyl biphenyl
630553
CCCCCC1CCC(CC1)C1=CC=C(C=C1)C1=CC=C(CC)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 145°C to 147°C
Quantity 1 g
IUPAC Name 1-ethyl-4-[4-(4-pentylcyclohexyl)phenyl]benzene
Formula Weight 334.54
Percent Purity 97%
Chemical Name or Material trans-4-Ethyl-4'-(4-n-pentylcyclohexyl)biphenyl
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'