Learn More
Thermo Scientific Chemicals Tolazoline hydrochloride, 99%
Catalog Number p-3758818
Click to view available options
Quantity:
50 g
250 g
Packaging:
Plastic bottle
59-97-2
C10H12N2·HCl
196.72
MFCD00012693
RHTNTTODYGNRSP-UHFFFAOYSA-N
tolazoline hydrochloride, tolazoline hcl, 2-benzyl-4,5-dihydro-1h-imidazole hydrochloride, arterodyl, tolavad, tolpal, priscoline hydrochloride, tolazoline chloride, imidaline hydrochloride, benzazoline hydrochloride
6048
2-benzyl-4,5-dihydro-1H-imidazole;hydrochloride
C1CN=C(N1)CC2=CC=CC=C2.Cl
59-97-2
C10H12N2·HCl
196.72
MFCD00012693
RHTNTTODYGNRSP-UHFFFAOYSA-N
tolazoline hydrochloride, tolazoline hcl, 2-benzyl-4,5-dihydro-1h-imidazole hydrochloride, arterodyl, tolavad, tolpal, priscoline hydrochloride, tolazoline chloride, imidaline hydrochloride, benzazoline hydrochloride
6048
2-benzyl-4,5-dihydro-1H-imidazole;hydrochloride
C1CN=C(N1)CC2=CC=CC=C2.Cl
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 172.0°C to 176.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 50 g
Merck Index 15, 9666
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in chloroform, freely soluble in alcohol, slightly soluble in ether and ethyl acetate
Formula Weight 196.72
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Tolazoline hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Tolazoline hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.