missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ TNFRSF17 (Human) Recombinant Protein
Human TNFRSF17 full-length ORF ( AAH58291, 1 a.a. - 184 a.a.) recombinant protein with GST-tag at N-terminal.
335.00€ - 508.00€
Specifications
Accession Number | AAH58291 |
---|---|
Gene ID (Entrez) | 608 |
Name | tumor necrosis factor receptor superfamily, member 17 |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16152191
|
Abnova™
H00000608-P01.10UG |
10 μg |
335.00€
10µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
16162191
|
Abnova™
H00000608-P01.25UG |
25 μg |
508.00€
25µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
Description
- Sequence: MLQMAGQCSQNEYFDSLLHACIPCQLRCSSNTPPLTCQRYCNASVTNSVKGTNAILWTCLGLSLIISLAVFVLMFLLRKISSEPLEDEFKNTGSGLLGMANIDLEKSRTGDEIILPRGLEYTVEECTCEDCIKSKPKVDSDHCFPLPAMEEGATILVTTKTNDYCKSLPAALSATEIEKSISAR
Specifications
AAH58291 | |
tumor necrosis factor receptor superfamily, member 17 | |
125% SDS-PAGE Stained with Coomassie Blue | |
BCM, BCMA, CD269 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
608 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TNFRSF17 | |
GST |