missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNF Rat anti-Mouse, BB700, Clone: MP6-XT22, BD Horizon™
Rat Monoclonal Antibody
Brand: BD Biosciences 566511
Additional Details : Weight : 0.00810kg
Description
TNF Monoclonal antibody specifically detects TNF in Mouse samples. It is validated for Flow Cytometry.Specifications
TNF | |
Monoclonal | |
0.2 mg/mL | |
Rat | |
RUO | |
Mouse | |
Purified |
Flow Cytometry | |
MP6-XT22 | |
Brilliant Blue 700 (BB700) | |
25 μg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG1 |