Learn More
Lovibond™ Tintometer™ CHECKIT™ Comparator, Silica LR Test, 0.25 to 4mg/L
Suitable for both mobile and static water analysis. Checkit™ Comparators are available with a number of different color scales and provide the basis for a comprehensive easy to to to use analysis system.
Brand:  Lovibond™ 147350
Additional Details : Weight : 0.46000kg Transport : UN number : 2967 Chem class : 8 Pack group : III
Product Code. 10653723
  • 75.20€ / Each
Estimated Shipment: 28-05-2024
Description
  • The Lovibond™ Checkit™ comparator is a compact, lightweight colorimetric unit
  • Suitable for both mobile and static water analysis
  • Available with a number of different color scales
  • Provides the basis for a comprehensive easy to to to use analysis system
  • Each Checkit™ Comparator Disc contains a continuous colour scale for exact color matching between standard and sample
  • Each Checkit™ Comparator Kit is supplied in a carry case with comparator, vials, discs, reagents and method instructions for all tests
  • The Silica disc supplied measures from 0.25 to 4mg/L with the following steps: 0.25 / 0.5 / 0.75 / 1.0 / 1.25 / 1.5 / 1.75 / 2.0 / 2.5 / 3.0 / 3.5 / 4.0
Specifications
Specifications
Water Quality Testing
Carry case with Comparator, Vials, Discs, reagents and method instructions for all tests
Silica LR Test
Checkit™ Comparator
0.25 to 4mg/L
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Testkit Checkit Comparator Siliciumdioxid LR 0,25
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P313-Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : sulphamic acid, sodium molybdate, tartaric acid, boric acid