Learn More
Ark-Plas Products™ Threaded Check Valve
Made from kynar. Ark-Plas Products™ Threaded Check Valve is made from a high-quality manufacturing process that helps to ensure a leak-proof seal. The valve is a zero-gravity valve that detects the slightest change in pressure or flow.
Brand:  Ark-Plas Products™ HEX40-DF0
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11891180
  • 127.40€ / Pack of 10
Estimated Shipment
04-05-2021
Add to basket
Description
Description
  • Made from Viton
  • Free-floating design
  • Zero gravity fitting
Specifications
Specifications
Natural
Kynar
1/8-27 NPT
3/16 in. valve