missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Fluka™ Thorin, For Spectrometric Determination, Honeywell™ Fluka™

for spectrophotometric det. of Li, Th, Zr

Brand:  Honeywell Fluka™ 89120-10G

Additional Details : CAS : 3688-92-4 Weight : 0.01000kg

Packaging Glass bottle
Quantity 10g

 View more versions of this product

Product Code. 15627380

  • 147.61€ / 10g


please call +351214253350
Add to basket

Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

3688-92-4
576.289
ZQMPNLOZJUWHSA-WTBAQZMLSA-L
6537679
C1=CC=C(C(=C1)NN=C2C3=C(C=C(C=C3)S(=O)(=O)[O-])C=C(C2=O)S(=O)(=O)[O-])[As](=O)(O)O.[Na+].[Na+]
C16H11AsN2Na2O10S2
MFCD00003888
thoron, thoron ol, 1-2-arsonophenylazo-2-naphthol-3,6-disulfonic acid disodium salt, apans, aspan, disodium 4e-4-2-arsonophenyl hydrazinylidene-3-oxonaphthalene-2,7-disulfonate, thoronol, o-3,6-disulfo-2-hydroxy-1-naphthyl-azo-benzenearsonic acid, 2-2-hydroxy-3,6-disulfo-1-naphthylazo phenylarsonic acid disodium salt, 2-3,6-disulfo-2-hydroxy-1-naphthylazo benzenearsonic acid disodium salt
disodium;(4E)-4-[(2-arsonophenyl)hydrazinylidene]-3-oxonaphthalene-2,7-disulfonate

Specifications

Thorin
Glass bottle
10g
3688-92-4
MFCD00003888
2957648
Soluble in water
C1=CC=C(C(=C1)NN=C2C3=C(C=C(C=C3)S(=O)(=O)[O-])C=C(C2=O)S(=O)(=O)[O-])[As](=O)(O)O.[Na+].[Na+]
576.289
576.3g/mol
Odorless
Red
6.1
C16H11AsN2Na2O10S2
UN3465
thoron, thoron ol, 1-2-arsonophenylazo-2-naphthol-3,6-disulfonic acid disodium salt, apans, aspan, disodium 4e-4-2-arsonophenyl hydrazinylidene-3-oxonaphthalene-2,7-disulfonate, thoronol, o-3,6-disulfo-2-hydroxy-1-naphthyl-azo-benzenearsonic acid, 2-2-hydroxy-3,6-disulfo-1-naphthylazo phenylarsonic acid disodium salt, 2-3,6-disulfo-2-hydroxy-1-naphthylazo benzenearsonic acid disodium salt
ZQMPNLOZJUWHSA-WTBAQZMLSA-L
disodium;(4E)-4-[(2-arsonophenyl)hydrazinylidene]-3-oxonaphthalene-2,7-disulfonate
6537679
Videos
Documents