Learn More
Thermo Scientific™ Thermo-Fast™ 96-Well Full-Skirted Plates
96-well full-skirted PCR automation compatible plate for use in PCR and qPCR applications.
113.00€ - 670.00€
Specifications
Certifications/Compliance ANSI; Certified free from human DNA, RNases, and DNases
For Use With (Application) PCR, qPCR applications
Volume (Metric) Well 200 μL
Well Color Blue
Material Polypropylene
View More Specs
Products 10
Product Code Brand Color No. per Pack Price Quantity & Availability  
Product Code Brand Color No. per Pack Price Quantity & Availability  
15504394 View Documents Thermo Scientific™
AB0800150
Clear 150 670.00€
Pack of 150 View alternative products
Save Up to 
»
EU Stock 3
Add to basket
10354273 View Documents Thermo Scientific™
AB0800B
Blue 25 119.00€
Pack of 25 View alternative products
Save Up to 
»
EU Stock 3
Add to basket
10039522 View Documents Thermo Scientific™
AB0800
Clear 25 120.00€
Pack of 25 View alternative products
Save Up to 
»
EU Stock 571
Add to basket
10069282 View Documents Thermo Scientific™
AB0800G
Green 25 119.00€
Pack of 25 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 04-10-2019
Add to basket
10161073 View Documents Thermo Scientific™
AB0800R
Red 25 119.00€
Pack of 25 View alternative products
Save Up to 
»
EU Stock 12
Add to basket
10170593 View Documents Thermo Scientific™
AB0800Y
Yellow 25 119.00€
Pack of 25 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 25-10-2019
Add to basket
10334513 View Documents Thermo Scientific™
AB0800K
Black 25 131.00€
Pack of 25 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 04-10-2019
Add to basket
10794404 View Documents Thermo Scientific™
AB0800W
White 1 131.00€
Pack of 25 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 01-10-2019
Add to basket
10708073 View Documents Thermo Scientific™
AB0800P
Purple 25 119.00€
Pack of 25 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 25-10-2019
Add to basket
15737486 View Documents Thermo Scientific™
AB0800
Clear 25 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
Description
Description
96-Well Full-Skirted Plates have a low-profile, thin well wall design and are compatible with PCR and qPCR applications. They feature an SBS footprint, and the plates are stackable for use in automated systems. These plates are available as SuperPlate, which are more rigid than standard plates, making it easier for robotic handling. The 96-Well Full-Skirted Plates feature a cut corner at the H1 location. These plates also have eight holes in the skirt, aiding in plate positioning and removal from the thermal cycler block. The full skirt surround provides an even surface for robotic arms to grip, making the plate suitable for use with automated systems.
The Thermo Scientific 96-Well Full-Skirted Plates have a low-profile, thin well wall design and are compatible with PCR and qPCR applications. They feature an SBS footprint, and the plates are stackable for use in automated systems. These plates are available as SuperPlate, which are more rigid than standard plates, making it easier for robotic handling. The 96-Well Full-Skirted Plates feature a cut corner at the H1 location. These plates also have eight holes in the skirt, aiding in plate positioning and removal from the thermal cycler block. The full skirt surround provides an even surface for robotic arms to grip, making the plate suitable for use with automated systems.
Highlights
  • ANSI:SBS footprint and stackable for use in automated systems
  • Low profile to reduce dead space and increase PCR efficiency
  • Available as SuperPlate, providing 4x more rigidity for superior robotic handling
  • 0.2 mL maximum well volume (when used with adhesive and heat seals)
  • Clear well for sample visibility
  • Sealable using Thermo-Mats, Adhesive Films and Foils, Heat Seals, and all Thermo Scientific Cap Strips
  • 8 holes in the skirt aid plate positioning and removal from the thermal cycler block
  • Raised rim design around each well enables secure sealing and safeguards against evaporation
  • Uniform, thin well walls deliver maximum and consistent heat transfer
  • Black lettered versions feature black alpha-numeric lettering to allow for easy well identification
  • White plates are optimized for qPCR by increasing sensitivity and reducing variability in fluorescence detection
  • Cut Corner: H1
Applications
  • PCR
  • qPCR/Real-Time PCR
Certifications
Certified free from human DNA, RNases, and DNases.
Specifications
Specifications
ANSI; Certified free from human DNA, RNases, and DNases
200 μL
Polypropylene
Full
PCR, qPCR applications
Blue
Yes
Without strengthing ribs