Learn More
Thermo Scientific™ High Purity Two Stage Brass Gas Regulators
Recommended for non-corrosive analytical and process applications requiring precise, stable, delivery pressure control
1010.00€ - 1400.00€
Specifications
Range | 0 to 200 psig |
---|
Includes: Diffusion-resistant, diaphragm packless outlet valve to maintain system purity
Product Code | Brand | Range | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Range | Type | Price | Quantity & Availability | ||||
10117984
|
Thermo Scientific™
60180-926 |
0 to 200 psig | CGA 590 with Combi Filter |
1370.26€
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||||
10448854
|
Thermo Scientific™
60180-908 |
0 to 200 psig | DIN 1 with Triple Filter |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
10046844
|
Thermo Scientific™
60180-906 |
0 to 200 psig | CGA 350 with Triple Filter |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
10065674
|
Thermo Scientific™
60180-920 |
0 to 200 psig | CGA 350 with Combi Filter |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
10544315
|
Thermo Scientific™
60180-902 |
0 to 200 psig | CGA 580 with Triple Filter |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
Description
- For non-corrosive analytical and process applications
- Includes a built-in Smart Purifier Base and Cartridge ensuring 99.99999% gas purity
- Specially designed, convoluted, stainless steel diaphragm provides goodregulating performance and maximum purity integrity without the needfor a soft seal
- Capable of withstanding an internal vacuum
- All regulators supplied with 1/8 in. stainless steel output adaptor
- Built in check valve allows for cylinder change without contaminating the gas line
Specifications
0 to 200 psig |