Learn More
Thermo Scientific™ Shelving Options and Accessories for Environmental Chambers and Incubators
Utilize shelving options and accessories to increase the flexibility of your Thermo Scientific™ Environmental Chamber or Incubator.
Brand:  Thermo Scientific™ 224139
Additional Details : Weight : 11.00000kg
 View more versions of this product
Product Code. 15503771
  • 251.00€ / Each
Estimated Shipment date
from Supplier 20-02-2019
Add to basket
Specifications
Specifications
Solid Stainless Steel Shelf and Channels
Stainless Steel
Models 3920, 3940 (3949), 3911 (3913) and 3960 (3961); Environmental chambers 3948, 3907 and 3962
Shelving
Videos
Cat No: 224161 -- Two per unit max., not for shakers; 68.0 kg load maximum with shelf fully inserted and stationary