Learn More
Abeba™ The Original 7204 Shoes
Animal fats, oil and fuel resistant. Sanitized™ treated.
Brand: Abeba™ 7204/39
Additional Details : Weight : 0.76300kg
Description
- CE EN ISO 20347:2012 OB FO SRC
- Smooth leather white with motif
- Slip resistance outsole sole
- Sanitized treated
- Heel strap foldable, adjustable
- Leather insole
- Discontinued model
- Heel cushion
Specifications
CE EN ISO 20347:2012 OB FO SRC | |
Leather | |
Slip-resistant | |
Animal fats, oil and fuel |
White | |
39 | |
7204 |