Learn More
Texwipe™ FoamWipe™ Polyurethane Foam Wipers
For ISO Class 7 to 8 (Class 10,000 to 100,000) applications
Brand: Texwipe™ TX704
Additional Details : Weight : 3.00000kg
Description
- Open-cell, 100 pore-per-inch polyurethane foam precision cut for low particle generation
- Excellent replacement for cellulose sponges in critical environments
- Can be used as a soft and nonabrasive work surface
Order Info
Basis weight: 105g/m2. Silicone-free and solvent-safe Bag-Within-A-Bag™ packaging.
Specifications
FoamWipe™ Wipes | |
22.9 cm | |
15.24 cm | |
105 | |
9 x 6 in. (22.8 x 15.2 cm) | |
0.125 in. |
9 in. | |
9 in. | |
Polyurethane | |
ISO 9001:2000, 13485:2003, 14001:2004 and OHSAS 18001:1999 | |
For use in cleanroom maintenance and assembly operations |