missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetramethylene sulfone, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 27.4°C to 27.8°C |
Color | Yellow |
Density | 1.2600g/mL |
Boiling Point | 285.0°C |
Flash Point | 165°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% min. (GC) |
Packaging | Glass bottle |
Refractive Index | 1.4820 to 1.4850 |
Quantity | 250 mL |
Show More |
Safety and Handling
Product Identifier
- Tetramethylene sulfone
- Danger
- Acute toxicity Category 4
- Reproductive toxicity Category 1B
- H302-Harmful if swallowed.
- H360-May damage fertility or the unborn child.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P201-Obtain special instructions before use.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'