missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetra-n-butylammonium difluorotriphenylstannate, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Can be regarded as a non-hygroscopic anhydrous equivalent of TBAF useful in the preparation of gem-difluorides and the alkylation of enol silyl ethers. For use in the fluorination of carbohydrates.
Solubility
Soluble in most organic solvents.
Notes
Hygroscopic. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Can be regarded as a non-hygroscopic anhydrous equivalent of TBAF useful in the preparation of gem-difluorides and the alkylation of enol silyl ethers. For use in the fluorination of carbohydrates.
Solubility
Soluble in most organic solvents.
Notes
Hygroscopic. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | 188°C to 191°C |
Quantity | 1 g |
UN Number | UN3146 |
Beilstein | 8668854 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in most organic solvents. |
Formula Weight | 630.5 |
Percent Purity | 97% |
Chemical Name or Material | Tetra-n-butylammonium difluorotriphenylstannate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'