missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testosterone-16,16,17-d3, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Testosterone-D3 (16,16,17-D3) |
| CAS | 77546-39-5 |
| Molecular Formula | C19 2H3 H25 O2 |
| Quantity | 0.01 g |
| Synonym | Androst-4-en-3-one-16,16,17-d3, 17-hydroxy-, (17β)-, (17β)-17-Hydroxyandrost-4-en-3-one-16,16,17-d3, 16,16,17-Testosterone-D3, Testosterone-16,16,17-D3, 19,19,19-Trideuteriotestosterone |
| InChI Formula | InChI=1 S/C19H28O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h11,14-17,21 H,3-10H2,1-2H3/t14-,15-,16-,17-,18-,19-/m0/s1/i6D2,17 D |
| SMILES | [2 H]C1([2 H])C[C@H]2 [C@@H]3 CCC4=CC(=O)CC[C@]4(C)[C@H]3 CC[C@]2(C)[C@@]1([2 H])O |
| IUPAC Name | (8 R,9 S,10 R,13 S,14 S,17 S)-16,16,17-trideuterio-17-hydroxy-10,13-dimethyl-1,2,6,7,8,9,11,12,14,15-decahydrocyclopenta[a]phenanthren-3-one |
| Molecular Weight (g/mol) | 291.44 |
| Formula Weight | 291.2278 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction