missing translation for 'onlineSavingsMsg'
Learn More

Binder™ Temperature Reinforced Rack

Manufactured to withstand extremely low temperatures.

Brand:  Binder™ 8012-0707

Additional Details : Weight : 1.50000kg

 View more versions of this product

Product Code. 15621866

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

  • For temperature range of −40° to −60°C
  • Set of 3

Specifications

Specifications

-40°C to -60°C
UF V 700
Reinforced Rack
Videos
Documents