Learn More
Thermo Scientific Chemicals TBE (5X), TRIS + borate + EDTA
Product Code. 15465709
Click to view available options
Quantity:
2 L
4 L
Unit Size:
2 L
4 L
Product Code. 15465709
Brand: Thermo Scientific Chemicals J63487.K3
Request Bulk or Custom Format
Edge
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material TBE (5X)
Physical Form Liquid
Quantity 2 L
MDL Number MFCD00236358
Packaging Plastic bottle
My Thermo Scientific brand TBE buffer has precipitated out of solution. What can I do?
Concentrations of 5X Tris Borate EDTA (TBE) buffer or greater may precipitate due to the nature of the product. This precipitation is a result of nucleation of salt crystals by dust particles or other insoluble materials. In some cases, we recommend heating the buffer to ~ 50°C to re-dissolve the TBE into solution. Alternately, to avoid precipitation of high concentration TBE buffers, after the solution is prepared, you can filter it through a 0.2 µm cellulose acetate or cellulose nitrate filter. These treatments have no deleterious effect on the buffering properties of TBE.