missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAS2R40 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567749
Additional Details : Weight : 0.01000kg
Description
Predicted to react with Mouse and Rat samples.
TAS2R40 Polyclonal antibody specifically detects TAS2R40 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
TAS2R40 | |
Polyclonal | |
Unconjugated | |
TAS2R40 | |
G protein-coupled receptor 60; GPR60; G-protein coupled receptor 60; MGC126844; MGC126846; mt2r33; RGD1562314; T2R18; T2r33; T2R40; T2R58; Tas2r144; TAS2R40; tas2r40 {ECO:0000250; Tas2r44; taste 2 receptor member 40; taste receptor type 2 member 18; Taste receptor type 2 member 40; taste receptor type 2 member 58; taste receptor, type 2, member 144; taste receptor, type 2, member 40; UniProtKB:P59535} | |
Rabbit | |
affinity chromatography | |
RUO | |
259286, 387515, 500101 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P59535, Q67ET2, Q7TQB8 | |
Tas2r144, TAS2R40 | |
Synthesized peptide derived from human TAS2R40. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |