Learn More
IBA Solutions For Life Science Tamra dUTP
A modified nucleoside-triphosphate (3’-modified oligos) that can be used for fluorescent labeling of DNA.
Brand:  IBA Solutions For Life Science 5-0618-002
Additional Details : Weight : 0.00050kg
 View more versions of this product
Product Code. 15623678
  • 473.00€ / 100nmol
Estimated Shipment date
from Supplier 29-07-2019
Add to basket
Description
Description
IBA has a nucleoside-triphosphate synthesis facility that offers an extensive range of triphosphates for many applications.
  • All products are double HPLC purified, desalted and well characterized by HPLC and 31P-NMR
  • They are also available as monophosphates and nucleosides
Specifications
Specifications
Yes
Fluorescent labeling of DNA
100 nmol
Yes
Nucleoside-triphosphate (3'-modified oligo)