Learn More
Thermo Scientific Chemicals TAE (50X), TRIS + acetate + EDTA
TAE (50X), TRIS + acetate + EDTA is a buffer composed of Tris, acetate, and EDTA components and is used in electrophoresis of nucleic acids in agarose and polyacrylamide gels.
118.00€ - 397.00€
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15476089
|
Thermo Scientific Chemicals
J63931.K2 |
1 L |
118.00€
1 L |
In Stock
Log in to see stock. |
|||||
15486089
|
Thermo Scientific Chemicals
J63931.K3 |
2 L |
209.00€
2 L |
In Stock
Log in to see stock. |
|||||
15496089
|
Thermo Scientific Chemicals
J63931.K7 |
4 L |
397.00€
4 L |
In Stock
Log in to see stock. |
|||||
Description
TAE (50X), TRIS + acetate + EDTA is used in electrophoresis of nucleic acids in agarose and polyacrylamide gel. It acts as a running buffer and as a gel preparation buffer. It is also useful for nucleic acid separations viz. DNA and RNA. Further, it is used for non-denaturing RNA agarose gel electrophoresis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• TAE (50X), TRIS + acetate + EDTA allows nucleic acid separation
Application
• This buffer has been used to prepare agarose and polyacrylamide gels for electrophoresis of nucleic acids
• It is used as a running and gel preparation buffer, and it is also used for non-denaturing RNA agarose gel electrophoresis
RUO – Research Use Only