Learn More
Sigma Laborzentrifugen™ Swing-out Rotors for Sigma™ Series 2 Centrifuges
Use these rotors with maximum load and at maximum speed for up to 50,000 cycles with low maintenance. Sigma™ Swing-out Rotors for Sigma™ Series 2 Centrifuges are ideal for use in medical and research laboratories.
Brand:  Sigma Laborzentrifugen™ 11030
Additional Details : Weight : 5.00000kg
 View more versions of this product
Product Code. 10036111
  • 650.00€ / Each
Estimated Shipment: 06-03-2023
Description
  • Swing-bucket rotors for sedimentation with smaller centrifugal forces of up to approximately 6000 x g
  • A higher yield may be obtained as it is possible to fill the centrifuge test tube completely
  • Sedimentation takes place in the middle of the bottom of the test tube enabling easy separation of the phases
  • Sigma supplies matching fixed-angle, swing-out and drum rotors for all Sigma centrifuges
  • Sigma rotors are precision-made from modern, certified high-performance materials with traceable batch numbers
  • Thanks to many years of experience and close contact with our customers, we are able to offer an extensive selection of rotors
  • Sigma rotors are characterized by extra capacity or higher maximum speed
  • Rotors can be operated continuously at maximum speed and +4°C in nearly all Sigma centrifuges
Specifications
Specifications
Swing-out Rotor
4,000 rpm
4
120 mL
Sigma 2-6, 2-6E, Buckets: 13031, 13033, 13034, 13035, 13037