Learn More
Fisherbrand™ SureOne™ Low Retention Non-Filtered Pipette Tips, 200 μL, Standard Length
GREENER_CHOICE
Product Code. 16460875 Shop All Fisherbrand Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Packaging:
Bulk
Hinged Rack
Reload
Unit Size:
Pack of 1000
Pack of 960
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Packaging unitSize
16460875 Non-sterile Bulk Pack of 1000
16470875 Non-sterile Hinged Rack Pack of 960
16490875 Sterile Hinged Rack Pack of 960
16480875 Non-sterile Reload Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Single Use Only
Product Code. 16460875
  • ONLINE EXCLUSIVE PRICE
    21.30€
    20.80€ / Pack of 1000
    Save 0.50€
    2% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Single Use Only
Fisherbrand™ SureOne™ Low Retention Pipette Tips successfully minimize the formation of liquid films and build up on tip walls during dispensing, which provides maximum sample recovery and reproducibility.
  • Low retention technology helps maximize sample recovery
  • Universal fit
  • Non-filtered
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Non-sterile
Color Natural
Rack Color Blue
Autoclavable Autoclavable
Length (English) 1.98 in.
Length (Metric) 51 mm
Certifications/Compliance COA
Clean Claims Certified Free of: Rnase/Dnase, DNA, Pyrogen, Lot Traceable
Storage Requirements Ambient
Features Low Retention Technology, Ultra Clear Resin
Tip Style Low Retention, Beveled, Conical
Packaging Bulk
Quantity 1000
Product Type Pipette Tip
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated No
Product Line SureOne
Material Polypropylene
Show More Show Less
As part of our commitment to continuous product improvement, your delivery may contain products with some minor color variation, but their form, fit and function will not be affected. Any questions contact your sales representative.