Learn More
Thermo Scientific™ SuperSignal™ Western Blotting Kits
Combines SuperSignal West Pico substrate with HRP-conjugated rabbit or mouse secondary antibodies for a convenient, easy-to-use Western blot detection kit.
360.00€ - 502.00€
| Product Code | Brand | Description | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Description | Price | Quantity & Availability | |||||
| 10219963 |
Thermo Scientific™
34081 |
SuperSignal West Pico Complete Mouse IgG Detection Kit |
502.00€ 1 set |
from Supplier 04-09-2019 |
10495695 |
Thermo Scientific™
34082 |
SuperSignal West Pico Mouse IgG Detection Kit |
360.00€ 1 set |
from Supplier 30-09-2019 |
10590814 |
Thermo Scientific™
34083 |
SuperSignal West Pico Rabbit IgG Detection Kit |
360.00€ 1 set |
from Supplier 04-09-2019 |
10721765 |
Thermo Scientific™
34084 |
SuperSignal West Pico Complete Rabbit IgG Detection Kit |
502.00€ 1 set |
from Supplier 04-09-2019 |
Description
All the components of these Western blotting kits are optimized for compatibility the SuperSignal West Pico Chemiluminescent Substrate and are available for the detection of mouse and rabbit secondary primary antibodies. All SuperSignal Western Blotting kits come with complete, detailed instructions for those new to the technique of Western Blotting and abbreviated protocols for the experienced user. The kits are also available in a standard format (substrate and optimized HRP-conjugates) or a complete format which also includes the Thermo Scientific SuperBlock Buffer and BupH Tris Buffered Saline.
Highlights:
Standard and complete kits – choose kits with just secondary antibody and chemiluminescent substrate or kits that also include blocking buffer and wash buffer
Mouse and rabbit kits – choose kits with goat anti-mouse IgG or goat anti-rabbit IgG secondary antibody conjugates
Instructions – complete, detailed manual with recommended antibody dilutions
Ordering Information:
Each kit provides sufficient reagents to process 50 miniblots or 4000cm2 of membrane.
Includes:
Standard Detection Kits Include: West Pico Substrate (luminol/enhancer solution and stable peroxide solution), HRP-conjugated anti-rabbit IgG, anti-mouse IgG, or biotin-binding protein. Complete Detection Kits also include SuperBlock Blocking Buffer and TBS wash buffer.