missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfamic Acid, ≥99%, Reagent Grade, Honeywell™
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Poly Bottle
Description
- ≥95% for general laboratory use.
Specifications
Appearance | Crystalline powder |
Color | Colorless |
Melting Point | 205°C |
Density | 2.1g/cm3 |
Packaging | Poly Bottle |
Quantity | 100 g |
Linear Formula | NH2SO3H |
UN Number | UN2967 |
Formula Weight | 97.09g/mol |
Percent Purity | ≥99% |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'