Learn More
Sampling Systems™ Sub Sampler Pro
Can sample from a set distance from the bottom of a container or alternatively at a mid point in a container or well. Sampling Systems™ Sub Sampler Pro is sometimes called a Bomb Sampler and it is used throughout the world for both chemical and environmental sampling.
Brand:  Sampling Systems™ 5072B-500
Additional Details : Weight : 2.88000kg
 View more versions of this product
Product Code. 15809361
  • 233.42€ / Each
Estimated Shipment: 27-10-2021
Add to basket
Description
Description
  • Lower Sub Sampler Pro until the plunger assembly reaches the bottom of the container
  • When the plunger assembly hits the bottom, it is pushed up allowing the sample to enter the device
  • When the sampler is raised the plunger will drop down and seal the sampler
  • Lower the Sub Sampler Pro into the liquid
  • At the required depth pull on the control line to raise the plunger
  • This will allow the liquid to enter the sampler
  • Release the control Line and the plunger will drop down and seal the sample
Specifications
Specifications
500mL
Stainless Steel
Sampler