Learn More
Sampling Systems™ SteriWare™ Disposable Tray
Sampling Systems™ SteriWare™ Disposable Tray is ideal for hygienic applications within hospitals and pharmaceutical factories.
Brand: Sampling Systems™ 8081P-3000
Additional Details : Weight : 29.36640kg
Description
- Moulded and packed in a Class 100,000 Cleanroom
- Individually bagged
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
- BSE/TSE free
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- No cross contamination
- No cleaning required
Specifications
Polypropylene | |
Yes | |
White |
3 L | |
Non-sterile | |
310 x 265 x 55 mm |