Learn More
Standard Buffer Phosphate, pH 6.881, NIST, Chem-Lab
For laboratory use, pH measurements, traceable to SRM from NIST
39.05€ - 248.00€
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11984024
|
Chem Lab
CL03.0601.1000 |
1 L |
39.05€
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
11918494
|
Chem Lab
CL03.0601.9010 |
10 L |
248.00€
10 L |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
Description
pH = 6.984 at 0°CpH = 6.951 at 5°C
pH = 6.923 at 10°C
pH = 6.900 at 15°C
pH = 6.881 at 20°C
pH = 6.865 at 25°C
pH = 6.853 at 30°C
pH = 6.838 at 40°C
pH = 6.833 at 50°C
pH = 6.836 at 60°C
pH = 6.845 at 70°C
pH = 6.859 at 80°C