missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Standard Buffer Borate, pH 9.225, NIST, Chem-Lab
Click to view available options
Quantity:
1 L
10 L
Description
pH = 9.464 at 0°C
pH = 9.395 at 5°C
pH = 9.332 at 10°C
pH = 9.276 at 15°C
pH = 9.225 at 20°C
pH = 9.180 at 25°C
pH = 9.139 at 30°C
pH = 9.068 at 40°C
pH = 9.011 at 50°C
pH = 8.962 at 60°C
pH = 8.921 at 70°C
pH = 8.885 at 80°C
pH = 9.395 at 5°C
pH = 9.332 at 10°C
pH = 9.276 at 15°C
pH = 9.225 at 20°C
pH = 9.180 at 25°C
pH = 9.139 at 30°C
pH = 9.068 at 40°C
pH = 9.011 at 50°C
pH = 8.962 at 60°C
pH = 8.921 at 70°C
pH = 8.885 at 80°C
Specifications
Density | 1.01g/mL |
pH | 9.225 |
Quantity | 1 L |
Packaging | PE |
Traceability to NIST | NIST traceable |
Formula Weight | 381.37g/mol |
Chemical Name or Material | Sodium tetraborate decahydrate solution |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'