Learn More
RUBIS Mini Tweezers
Mini Tweezers - Antimagnetic/Anti-acid SS
Brand: RUBIS 2M
Additional Details : Weight : 0.01000kg
Description
- Mini Tweezers
- High Precision Tweezers
- 100% Made in Switzerland
- Anti-acid/Antimagnetic Stainless Steel
- Perfect alignment
- Very effective in narrow spaces
- Excellent for use under microscope
- Reliable
- OAL: 70 mm (2.75 in.)
Specifications
Pointed, Fine | |
Anti-acid, Anti-magnetic Stainless Steel | |
Mini Tweezers | |
Each |
2.76 in. | |
70 mm | |
Each |