Learn More
Sampling Systems™ Stainless Steel Mini Pots with Lids
Constructed of high quality stainless steel for durability. Sampling Systems™ Stainless Steel Mini Pots with Lids have a crevice free interior and are widely used in pharmaceutical and chemical laboratories.
41.80€ - 451.00€
Specifications
Product Type | Sample Jar |
---|---|
Material | Stainless Steel (316L) |
Closure Type | Threaded Cap |
Includes | Lid and body |
Product Code | Brand | Capacity (Metric) | Diameter (Metric) | Height (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Diameter (Metric) | Height (Metric) | Price | Quantity & Availability | |||
15819661
|
Sampling Systems™
A426-100 |
100 mL | 52 mm | 73 mm |
109.00€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||
15829661
|
Sampling Systems™
A426-1000 |
1000 mL | 102 mm | 154 mm |
451.00€
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
15839661
|
Sampling Systems™
A426-25 |
25 mL | 41 mm | 36 mm |
69.30€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||
15849661
|
Sampling Systems™
A426-250 |
250 mL | 77 mm | 73 mm |
197.00€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||
15859661
|
Sampling Systems™
A426-50 |
50 mL | 42 mm | 53 mm |
76.50€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||
15869661
|
Sampling Systems™
A426-500 |
500 mL | 102 mm | 78 mm |
301.00€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||
15879661
|
Sampling Systems™
A426-7 |
7 mL | 19 mm | 45 mm |
41.80€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||
15889661
|
Sampling Systems™
A426-750 |
750 mL | 102 mm | 116 mm |
318.00€
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
Description
- Available in a large range of sizes
- Screw on lid
- Satin finish
- Multi purpose small volume containers
- 316L Stainless Steel
- Crevice free interior
- Silicone O Ring available for improved sealing
Specifications
Sample Jar | |
Stainless Steel (316L) | |
Threaded Cap | |
Lid and body |