Learn More
Linker Industrietechnik™ Stainless Steel Collection Pan For 400 dia. x 70mm Internal Height Seives
Brand:  Linker Industrietechnik™ 554.070.200-905
Additional Details : Weight : 1.42000kg
Product Code. 11702187
  • 346.62€ / Each
Estimated Shipment: 05-06-2024
Specifications
For 400 dia. x 70mm Internal Height seives
1Each
Stainless Steel
400 mm
Collecting Pan