Learn More
Linker Industrietechnik™ Stainless Steel Collection Pan For 150 dia. x 50mm Internal Height Seives
Brand: Linker Industrietechnik™ 551.550.200-905
This item is not returnable.
View return policy
Specifications
1Each | |
Stainless Steel | |
150 mm |
Collecting Pan | |
For 150 dia. x 50mm Internal Height seives |