Learn More
SPEX™ SamplePrep Mid-Size Grinding Vial Set
Ideal for grinding samples such as plant and animal tissue, seeds, polymers, pharmaceuticals, textiles, hair, and bones
829.43€ - 1042.20€
Includes: One 6881P Mid-Size Stainless-steel Impactor, two 6881E Mid-Size Stainless-steel End Plugs, and one pack of 6881C4 Mid-Size Polycarbonate Center Cylinders
Requires: 886 Mid-Size Grinding Vial Adapter for grinding in the 6870 Large Freezer/Mill and 6884 Mid-Size Grinding Vial Adapter for Large Extractor/Vial Opener.
| Product Code | Brand | For Use With (Equipment) | Product Type | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | For Use With (Equipment) | Product Type | Price | Quantity & Availability | ||||
| 12898340 |
SPEX™ SamplePrep
6881 |
Large extractor/vial opener conjunction with mid size grinding vial adapter | Grinding Vial Set |
912.60€ 1 set |
|
15340482 |
SPEX™ SamplePrep
6881C20 |
Use with 6881 Vial Set | Cylinder |
829.43€ Pack of 20 |
|
15370482 |
SPEX™ SamplePrep
6883 |
Mid-Size Cr-free Vial |
1042.20€ 1 set |
|
Description
Mid-Size Grinding Vial Set contains everything necessary to make one complete grinding vial plus three spare polycarbonatec enter cylinders.
- Grinds one sample at a time in the 6870 Large Freezer/Mill™ when used in conjunction with the 6886 Mid-Size Grinding Vial Adapter
- Sample grinding capacity: 3 to 25g (25mL)
- Withstands liquid nitrogen temperatures
- Closed grinding vial prevents sample cross-contamination, retains volatiles, and controls hazardous or critical samples
- Transparent plastic center cylinder allows visual checking of the grinding progress
- Stainless-steel end plugs and impactor are autoclavable and reusable
- Polycarbonate center cylinders are disposable or may be cleaned with hot water and detergent or RNase/DNase-free solution
- Compatible with 6804 Large Extractor/Vial Opener when used in conjunction with 6884 Mid-Size Grinding Vial Adapter