Learn More
SPEX™ SamplePrep Mid-Size Grinding Vial Set
Ideal for grinding samples such as plant and animal tissue, seeds, polymers, pharmaceuticals, textiles, hair, and bones
829.43€ - 1042.20€
Includes: One 6881P Mid-Size Stainless-steel Impactor, two 6881E Mid-Size Stainless-steel End Plugs, and one pack of 6881C4 Mid-Size Polycarbonate Center Cylinders
Requires: 886 Mid-Size Grinding Vial Adapter for grinding in the 6870 Large Freezer/Mill and 6884 Mid-Size Grinding Vial Adapter for Large Extractor/Vial Opener.
Products 3
Product Code Brand For Use With (Equipment) Product Type Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Product Type Price Quantity & Availability  
12898340 View Documents SPEX™ SamplePrep
6881
Large extractor/vial opener conjunction with mid size grinding vial adapter Grinding Vial Set 912.60€
1 set View alternative products
Save Up to 
»
Estimated Shipment
09-10-2019
Add to basket
15340482 View Documents SPEX™ SamplePrep
6881C20
Use with 6881 Vial Set Cylinder 829.43€
Pack of 20 View alternative products
Save Up to 
»
Estimated Shipment
09-10-2019
Add to basket
15370482 View Documents SPEX™ SamplePrep
6883
Mid-Size Cr-free Vial 1042.20€
1 set View alternative products
Save Up to 
»
Estimated Shipment
09-10-2019
Add to basket
Description
Description
Mid-Size Grinding Vial Set contains everything necessary to make one complete grinding vial plus three spare polycarbonatec enter cylinders.
  • Grinds one sample at a time in the 6870 Large Freezer/Mill™ when used in conjunction with the 6886 Mid-Size Grinding Vial Adapter
  • Sample grinding capacity: 3 to 25g (25mL)
  • Withstands liquid nitrogen temperatures
  • Closed grinding vial prevents sample cross-contamination, retains volatiles, and controls hazardous or critical samples
  • Transparent plastic center cylinder allows visual checking of the grinding progress
  • Stainless-steel end plugs and impactor are autoclavable and reusable
  • Polycarbonate center cylinders are disposable or may be cleaned with hot water and detergent or RNase/DNase-free solution
  • Compatible with 6804 Large Extractor/Vial Opener when used in conjunction with 6884 Mid-Size Grinding Vial Adapter