Learn More
Spectrum™ Spectrum™ Dialysis Tubing Closures: Spectra/Por™ Standard Type
Consist of a single-piece, hinged clamp with sealing ridges that securely lock over the dialysis tubing to provide a leak-proof seal
153.00€ - 220.00€
Product Code | Brand | Color | Width (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Width (Metric) | Price | Quantity & Availability | ||||
10012284
|
Spectrum™
142936 |
Blue | 35 mm |
188.00€
Pack of 10 |
Estimated Shipment: 16-04-2024
Log in to see stock. |
||||
10002244
|
Spectrum™
132734 |
Orange | 12 mm |
153.00€
Pack of 10 |
Estimated Shipment: 16-04-2024
Log in to see stock. |
||||
11770753
|
Spectrum™
142736 |
White | 35 mm |
188.00€
Pack of 10 |
Estimated Shipment: 16-04-2024
Log in to see stock. |
||||
11425969
|
Spectrum™
132735 |
Orange | 23 mm |
200.00€
Pack of 10 |
Estimated Shipment: 16-04-2024
Log in to see stock. |
||||
11445969
|
Spectrum™
132738 |
Orange | 75 mm |
219.00€
Pack of 10 |
Estimated Shipment: 16-04-2024
Log in to see stock. |
||||
11348280
|
Spectrum™
142836 |
Green | 35 mm |
188.00€
Pack of 10 |
Estimated Shipment: 10-05-2024
Log in to see stock. |
||||
11302613
|
Spectrum™
132737 |
Orange | 55 mm |
220.00€
Pack of 10 |
Estimated Shipment: 18-04-2024
Log in to see stock. |
||||
11435969
|
Spectrum™
132736 |
Orange | 35 mm |
206.00€
Pack of 10 |
Estimated Shipment: 16-04-2024
Log in to see stock. |
||||
Description
Consist of a single-piece, hinged clamp with sealing ridges that securely lock over the dialysis tubing to provide a leak-proof seal
Spectra/Por Standard Closure Type:
- Made from polypropylene to aid in sample buoyancy
- Maintains sample vertical orientation when used with a Spectra/Por Weighted Closure.
- 5 sealing width sizes: 12mm, 23mm, 35mm, 55mm and 75mm
- 4 colors available for easier sample identification
- Autoclavable
- 10/pkg; Use with RC dialysis tubing only. Not for use with CE tubing.
Specifications
Autoclavable |