missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Fluka™ Sodium Thiocyanate, Honeywell Fluka™

ACS Reagent,≥98.0%

Brand:  Honeywell Fluka™ 251410-2.5KG

Additional Details : CAS : 540-72-7 Weight : 2.50000kg

Quantity 2.5kg

 View more versions of this product

Product Code. 15622740

  • 505.17€ / 2.50kg


EU Stock 5
Add to basket

Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

540-72-7
81.068
VGTPCRGMBIAPIM-UHFFFAOYSA-M
516871
sodium;thiocyanate
CNNaS
MFCD00011123
sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased
CHEBI:30952
C(#N)[S-].[Na+]

Specifications

Sodium Thiocyanate
CNNaS
MFCD00011123
3594965
VGTPCRGMBIAPIM-UHFFFAOYSA-M
sodium;thiocyanate
516871
81.07g/mol
ACS Reagent
2.5kg
540-72-7
NaSCN
NONH for all modes of transport
sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased
C(#N)[S-].[Na+]
81.068
CHEBI:30952
≥98%
Soluble in water, 1,000 g/L at 20°C
Videos
SDS Documents