missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiocyanate, ≥98.0% (on dried basis, AT), Honeywell™
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Poly bottle
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications
Quantity | 250 g |
Linear Formula | NaSCN |
UN Number | NONH for all modes of transport |
Beilstein | 3594965 |
IUPAC Name | sodium;thiocyanate |
Formula Weight | 81.07g/mol |
Percent Purity | ≥98.0% (on dried basis, AT) |
Packaging | Poly bottle |
Chemical Name or Material | Sodium thiocyanate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'