Learn More
Chem Lab™ Sodium Standard Solution (Plasma HIQU)
Brand:  Chem Lab™ CL0114010100
Additional Details : Weight : 0.12750kg Transport : UN number : 3264 Chem class : 8 Pack group : III
Quantity 100mL
 View more versions of this product
Product Code. 11909102
  • 68.30€ / 100mL
Estimated Shipment
12-03-2021
Specifications
1000μg/mL
1,02g/mL
100mL