missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Peroxide, purum p.a, ≥95%, Honeywell Fluka™
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Melting Point | 300°C |
Color | White |
Physical Form | Crystals |
Quantity | 250 g |
UN Number | UN1504 |
Solubility Information | 106.7g/L at 20°C |
Formula Weight | 77.98g/mol |
Percent Purity | ≥95% |
Packaging | Poly bottle |
Chemical Name or Material | Sodium peroxide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'