missing translation for 'onlineSavingsMsg'
Learn More

Sodium Peroxide, purum p.a, ≥95%, Honeywell Fluka™

Catalog Number 15636860
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly bottle
This item is not returnable. View return policy
1313-60-6
Na2O2
77.978
MFCD00003497
PFUVRDFDKPNGAV-UHFFFAOYSA-N
sodium peroxide, disodium peroxide, solozone, natriumsuperoxid, sodium dioxide, disodium dioxide, natrium peroxydatum, sodium oxide, per, flocool 180, sodium oxide
14803
disodium;peroxide
[O-][O-].[Na+].[Na+]
This item is not returnable. View return policy
1313-60-6
Na2O2
77.978
MFCD00003497
PFUVRDFDKPNGAV-UHFFFAOYSA-N
sodium peroxide, disodium peroxide, solozone, natriumsuperoxid, sodium dioxide, disodium dioxide, natrium peroxydatum, sodium oxide, per, flocool 180, sodium oxide
14803
disodium;peroxide
[O-][O-].[Na+].[Na+]

for Wurzschmitt-decomposition, ACS reagent, beads (small),≥95%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Melting Point 300°C
Color White
Physical Form Crystals
Quantity 250 g
UN Number UN1504
Solubility Information 106.7g/L at 20°C
Formula Weight 77.98g/mol
Percent Purity ≥95%
Packaging Poly bottle
Chemical Name or Material Sodium peroxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'