missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Metabisulfite, Certified AR for Analysis, Fisher Chemical™
25.13 EUR valid until 2024-03-30
Use promo code "21741" to get your promotional price.
Use promo code "21741" to get your promotional price.
Click to view available options
Quantity:
500 g
1 kg
Description
Sodium Metabisulfite, Certified AR for Analysis
Specifications
Melting Point | 150°C |
Color | White |
pH | 4 to 6 |
Physical Form | Powder/Solid |
Quantity | 500 g |
Formula Weight | 190.1g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'