missing translation for 'onlineSavingsMsg'
Learn More

Sodium Iodide, purum p.a, ≥99.0% (AT), Honeywell™

Catalog Number p-7120808
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly bottle
This item is not returnable. View return policy
7681-82-5
INa
149.894
MFCD00003532
FVAUCKIRQBBSSJ-UHFFFAOYSA-M
sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech
5238
CHEBI:33167
sodium;iodide
[Na+].[I-]
This item is not returnable. View return policy
7681-82-5
INa
149.894
MFCD00003532
FVAUCKIRQBBSSJ-UHFFFAOYSA-M
sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech
5238
CHEBI:33167
sodium;iodide
[Na+].[I-]

Purum p.a.,≥99.0% (AT)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY
Boiling Point 1304°C (1013hPa)
Melting Point 662°C
Color Colorless
Physical Form Solid
Quantity 250 g
Linear Formula NaI
UN Number UN3077
Solubility Information 1.793g/L (at 20°C)
Formula Weight 149.89g/mol
Percent Purity ≥99.0% (AT)
Odor Odorless
Packaging Poly bottle
Density Vapor Density: >1 (vs air)
Chemical Name or Material Sodium iodide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'