missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloride, Free-flowing, Reagent Grade, anhydrous, ≥99%, Honeywell Fluka™

Catalog Number 15627120
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
12 kg
25 kg
Packaging:
Fiberboard Box
Poly Bottle
This item is not returnable. View return policy
7647-14-5
ClNa
58.44
MFCD00003477
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
5234
CHEBI:26710
[Na+].[Cl-]
This item is not returnable. View return policy
7647-14-5
ClNa
58.44
MFCD00003477
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
5234
CHEBI:26710
[Na+].[Cl-]

Free-flowing, Reagent Grade, anhydrous,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Boiling Point 1,465°C
Melting Point 801°C (literature)
CAS Max % ≤100.0000%
Quantity 25 kg
Linear Formula NaCl
Beilstein 3534976
IUPAC Name sodium chloride
Formula Weight 58.44
Percent Purity ≥99%
Grade Reagent
Packaging Fiberboard Box
Density 2.160 g/cm3
Chemical Name or Material Sodium chloride, Free-flowing, Reagent Grade, anhydrous, ≥99%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'