missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium carbonate concentrate, For 1L standard solution, 0.05 M Na2CO3 (0.1N), Honeywell Fluka™
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
CAS Min % | ≥70.0000%, ≥30.0000% |
CAS Max % | <90.0000%, <50.0000% |
Physical Form | Concentrate |
Quantity | 6 x 1 Ea. |
Linear Formula | Na2CO3 |
UN Number | NONH for all modes of transport |
Beilstein | 4154566 |
Formula Weight | 105.99 |
Packaging | Ampoule |
Chemical Name or Material | Sodium carbonate concentrate, For 1L standard solution, 0.05 M Na2CO3 (0.1N) |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'